Chloramphenicol

Supplier: IBI Scientific

Synonyms: Chloromycetin®(R,R)-ChloramphenicolTega-Cetin

IB02080
IB02080EA 55.1 USD
IB02080
Chloramphenicol
Chloramphenicol

Chloramphenicol is used in the amplification of plasmids and colony hybridizations


Refrigerated shipping and storage is required for this product.


Chlroamphenicol is used at the following concentrations: Stringent plasmids (25µg/ml) and Relaxed Plasmids (170µg/ml). Antibiotic inhibition/sensitivity assay, Crystallinity and Chromatographic Purity: PASS.

Formula: Cl₂CHCONHCH(CH₂OH)CH(OH)C₆H₄NO₂
MW: 323.13 g/mol
MDL Number: MFCD00078159
CAS Number: 56-75-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR