West Condenser, No Hold Up, Ace Glass Incorporated
Supplier: Ace Glass
9297-05 9297-09 9297-106 9297-110
89074-332EA 177.12 USD
89074-332 89074-334 89074-336 89074-338
West Condenser, No Hold Up, Ace Glass Incorporated
Condensers
West Condenser, "No Hold-Up", with standard taper outer joint at top and standard taper inner joint at bottom
Jacket length is 200mm. Fabricated from borosilicate glass.
Ordering information: Available with either hose barbs or "Ace-Safe" connectors on inlet/outlet.