Condenser, Friedrichs, 200 mm, Ace Glass Incorporated
Supplier: Ace Glass
5969-10
89053-992EA 684.68 USD
89053-992
Condenser, Friedrichs, 200 mm, Ace Glass Incorporated
Condensers
This Friedrichs condenser is reflux controlled by regulating the jacket temperature
Provides a compact, spiral path for condensation. Bottom is a standard taper 24/40 inner joint. Jacket length is 200 mm. Use with 3/8-inch I.D. tubing, size D hose connections.