'A' Assembly for Rotary Evaporators, Ace Glass Incorporated
Supplier: Ace Glass
3950-10 3950-07
89083-104EA 1511.38 USD
89083-104 89083-108
'A' Assembly for Rotary Evaporators, Ace Glass Incorporated
Condensers
Complete "A" Assembly glassware set for many popular bench-scale rotary evaporators
Fabricated from borosilicate glass. Fits models 200/205 and Series 114-144. Available poly coated for safety, or uncoated. Includes the main glass condenser body with inner coil for angled installation, and heavy wall 1L receiving flask, The bottom condenser joint is spherical [SJ] 35/20 ball joint. The included round-bottom receiving flask has a [SJ] 35/20 socket joint. Heavy wall flasks feature walls that are approximately 30% heavier than standard flasks.
Ordering information: Includes 89060-600 o-ring, 89059-774 GL thread caps, 89059-810 angled hose connectors, and 89060-258 stainless steel pinch clamp. The bottom joint is spherical [SJ] 35/20.