Nalgene® 65 Braided Platinum-Cured Silicone Tubing, Thermo Scientific
Supplier: Thermo Fisher Scientific
8061-3070 8061-3130 8061-3190
63015-356CS 1695.85 USD
63015-356 63015-358 63015-360
Nalgene® 65 Braided Platinum-Cured Silicone Tubing, Thermo Scientific
Tubing Transfer Tubing
Tubing is durable, tasteless, odorless, non-cytotoxic, non-pyrogenic, and non-hemolytic.
  • Autoclavable
  • Also be sterilized by gamma radiation
This high-purity tubing is designed for a variety of pump and transfer applications including pharmaceutical, laboratory, bioprocess manufacturing and food beverage applications.