Mammalian GenLysate™ Human Total Protein Lysates, G-Biosciences
Supplier: G-Biosciences
NLH-01 NLH-09 NLH-17 NLH-19 SLH-0210 NLH-22 SLH-087 NLH-02 NLH-06 NLH-08 NLH-11 NLH-13 NLH-20 SLH-021 SLH-0211 SLH-024 SLH-025 SLH-027 SLH-029 SLH-081 SLH-082 SLH-089 SLH-090 NLH-03 NLH-04 NLH-15 SLH-023 SLH-026 SLH-028 SLH-088 NLH-12 NLH-18 NLH-05 NLH-14 NLH-16 NLH-21 SLH-0212 SLH-0213
82021-728EA 220.24 USD
82021-728 82021-760 82021-788 82021-796 82022-076 95029-322 95029-330 82021-732 82021-748 82021-756 82021-764 82021-772 82021-800 82022-074 82022-080 82022-100 82022-104 82022-112 82022-120 82022-124 82022-128 82022-136 95029-332 82021-736 82021-740 82021-752 82021-780 82022-096 82022-108 82022-116 82022-132 82021-768 82021-792 82021-744 82021-776 82021-784 82021-804 82022-084 82022-088
Mammalian GenLysate™ Human Total Protein Lysates, G-Biosciences
Cells
G-Biosciences GenLysate™ total protein lysates are ready to use for SDS-PAGE and Western blotting purposes.
  • Offered as a 150 µg lyophilized protein per vial
  • Ready for use
  • Wide variety of species and tissues
G-Biosciences GenLysates™ are total protein tissue and cell lysates that are extracted from the tissue or cell of interest
The total proteins are being extracted from whole tissue or cells following the method of Laemmli. The proteins are extracted in a denaturing buffer containing 62 mM Tris HCl and 2% SDS with G-Biosciences Protease ARREST™ protease inhibitor cocktail. This minimizes proteolytic damage to the proteins.
G-Biosciences GenLysate™ total protein lysates are offered as a 150 µg lyophilized protein per vial and, following reconstitution in DI water, are ready for use. G-Biosciences offers GenLysate™ in a wide variety of different species and tissues. Species include: human (both normal tissue and tumor tissue lysates available), mouse, rat, macaque (primate), rabbit, pig, plant-related, bacteria, insect, bird, frog, fish, marsupial, earthworm, and yeast.
To inhibit proteolysis, the GenLysates™ are prepared in a lysis buffer containing a complete protease inhibitor cocktail.