Syringe Rack, Hamilton Company
Supplier: Hamilton
204880
89168-730EA 502.63 USD
89168-730
Syringe Rack, Hamilton Company
Racks Syringe Racks
Hamilton Company offers the most comprehensive selection of standard and custom syringes in the industry
The Hamilton brushed stainless steel syringe rack keeps your workplace tidy. Store up to five syringes from the 700 and 7000 Microliter™ or 1700 Gastight® series. The plastic lip on the syringe rack is designed to keep syringes from making contact with the rack or other syringes and to prevent sample contamination. The syringe rack can be used as a free standing or wall model.
Each is manufactured to achieve the highest level of accuracy and precision possible.