Pentamethyldisiloxane 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L16577-06
L16577-14
AAAL16577-06EA
75.9
USD
AAAL16577-06
AAAL16577-14
Pentamethyldisiloxane 95%
Pentamethyldisiloxane
Purity: 95%
CAS No.: 1438-82-0
Molecular Formula: (CH3)3SiOSiH(CH3)2
Formula Weight: 148.35
Flash Point: -19°(-2°F)
Refractive Index: 1.3740
Boiling Point: 86-87°
Beilstein Reference: 1734568
UN No.: 1993
MDL No.: MFCD00053658
Formula:
C₅H₁₆OSi₂ Boiling Pt: 86…87 °C Density: 0.758 Flash Pt: –19 °C (–2 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00053658 CAS Number: 1438-82-0 EINECS: 215-873-5 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...