Sodium 99%, sticks in mineral oil
Supplier: Thermo Scientific Chemicals
|
Danger
|
L13285-36
L13285-36
AAAL13285-36EA
149.69
USD
AAAL13285-36
CAAAAL13285-36
Sodium 99%, sticks in mineral oil
Sodium
Formula:
Na MW: 22.99 g/mol Boiling Pt: 881 °C Melting Pt: 98 °C Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00085307 CAS Number: 7440-23-5 EINECS: 231-132-9 UN: 1428 ADR: "4.3",I Merck Index: 13,08641 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...