1-Octadecene tech. 90%, Technical Grade

Supplier: Thermo Scientific Chemicals
Danger

L11004-AU L11004-AK
AAAL11004-AUEA 68.14 USD
AAAL11004-AU AAAL11004-AK
1-Octadecene tech. 90%, Technical Grade
1-Octadecene

Purity: 90% CAS No.: 112-88-9 Molecular Formula: CH3(CH2)15CH=CH2 Formula Weight: 252.48 Flash Point: 154°(309°F) Refractive Index: 1.4440 Melting Point: 15-17° Boiling Point: 178-179°/15mm Beilstein Reference: 1761067 MDL No.: MFCD00009003

Formula: C₁₈H₃₆
MW: 252.48 g/mol
Boiling Pt: 315
Melting Pt: 17.5
Density: 0.789
Flash Pt: 154 °C (309 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009003
CAS Number: 112-88-9
EINECS: 204-012-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR