2-Chlorobenzenesulfonamide 98%
Supplier: Thermo Scientific Chemicals
L09082-03
L09082-06
AAAL09082-03EA
25.9
USD
AAAL09082-03
AAAL09082-06
2-Chlorobenzenesulfonamide 98%
2-Chlorobenzenesulphonamide
Purity: 98%
CAS No.: 6961-82-6
Molecular Formula: C6H6ClNO2S
Formula Weight: 191.64
Melting Point: 189-191°
Beilstein Reference: 2937540
Formula:
C₆H₆ClNO₂S Melting Pt: 189…191 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051974 CAS Number: 6961-82-6 EINECS: 230-156-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...