n-Heptacosane 97%

Supplier: Thermo Scientific Chemicals

Synonyms: Heptacosane

L07796-03 L07796-03 L07796-MD L07796-MD
AAAL07796-03EA 158 USD
CAAAAL07796-03 AAAL07796-03 CAAAAL07796-MD AAAL07796-MD
n-Heptacosane 97%
n-Heptacosane
Formula: H₃C(CH₂)₂₅CH₃
MW: 380.74 g/mol
Boiling Pt: 270 °C/15 mm
Melting Pt: 57…60 °C
Density: 0.7796 g/cm³ (60 °C)
Flash Pt: 120 °C(248 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009862
CAS Number: 593-49-7
EINECS: 209-792-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR