1-Ethylpropylamine 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
3-Pentylamine, 3-Aminopentane, Pentan-3-amine
L02250-22
L02250-14
L02250-06
AAAL02250-22EA
292
USD
AAAL02250-22
AAAL02250-14
AAL02250-06
1-Ethylpropylamine 98%
1-Ethylpropylamine
Purity: 98%
CAS No.: 616-24-0
Molecular Formula: C5H13N
Formula Weight: 87.17
Flash Point: 2°(35°F)
Refractive Index: 1.4055
Odor: Amine-like
Boiling Point: 89-91°
Beilstein Reference: 635658
UN No.: 1106
MDL No.: MFCD00008096
Formula:
C₅H₁₃N MW: 87.16 g/mol Boiling Pt: 90 Density: 0.748 Flash Pt: 2 °C (35 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008096 CAS Number: 616-24-0 EINECS: 210-471-6 UN: 1106 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...