4-Amino-o-anisic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Amino-2-methoxybenzenecarboxylic acid

H27419-14 H27419-06
AAH27419-14EA 69.7 USD
AAH27419-14 AAH27419-06
4-Amino-o-anisic acid 97%
4-Amino-o-anisic acid

Purity: 97% CAS No.: 2486-80-8 Molecular Formula: C8H9NO3 Formula Weight: 167.16 Melting Point: ca 150° dec. MDL No.: MFCD00114479

Formula: C₈H₉NO₃
MW: 167.16
Melting Pt: ∼ 150 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00114479
CAS Number: 2486-80-8
EINECS: 219-632-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR