2-Amino-4-fluorophenol 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-Fluoro-2-hydroxyaniline

H26165-06 H26165-03
AAH26165-06EA 188 USD
AAH26165-06 AAH26165-03
2-Amino-4-fluorophenol 97%
2-Amino-4-fluorophenol

Purity: 97% CAS No.: 399-97-3 Molecular Formula: C6H6FNO Formula Weight: 127.12 Melting Point: 138-141° MDL No.: MFCD00077451

Formula: C₆H₆FNO
Melting Pt: 138…141 °C
Storage Temperature: Ambient
MDL Number: MFCD00077451
CAS Number: 399-97-3
EINECS: None

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR