ω,ω-Difluoroacetophenone 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H25750-MD
H25750-03
H25750-06
AAH25750-MDEA
69.5
USD
AAH25750-MD
AAH25750-03
AAH25750-06
ω,ω-Difluoroacetophenone 95%
ω,ω-Difluoroacetophenone
Purity: 95%
CAS No.: 395-01-7
Molecular Formula: C8H6F2O
Formula Weight: 156.13
Refractive Index: 1.4955
Odor: Characteristic
Boiling Point: 53-54°/12mm
MDL No.: MFCD06246879
Formula:
C₈H₆F₂O Boiling Pt: 53…54 °C/12 mm Melting Pt: 48…50 °C Storage Temperature: Ambient |
MDL Number:
MFCD06246879 CAS Number: 395-01-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...