4-Chloro-3-iodophenol 98%

Supplier: Thermo Scientific Chemicals
Warning

B25547-09 B25547-03
AAB25547-03EA 42.89 USD
AAB25547-03 AAB25547-09
4-Chloro-3-iodophenol 98%
4-Chloro-3-iodophenol
Formula: C₆H₄ClIO
MW: 254.45 g/mol
Melting Pt: 75…77 °C
Storage Temperature: Ambient
MDL Number: MFCD00070769
CAS Number: 202982-72-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR