Congo red indicator
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Disodium 3,3'-[[1,1'-biphenyl]-4,4'-diylbis(azo)]bis(4-aminonaphthalene-1-sulphonate), Sodium 3,3'-([1,1'-biphenyl]-4,4'-diylbis(diazene-2,1-diyl))bis(4-aminonaphthalene-1-sulphonate), Disodium 3,3'-[[1,1'-biphenyl]-4,4'-diylbis(azo)]bis(4-aminonaphthalene-1-sulfonate), CI 22120, Sodium 3,3'-([1,1'-biphenyl]-4,4'-diylbis(diazene-2,1-diyl))bis(4-aminonaphthalene-1-sulfonate), Direct Red 28
CAS No.: 573-58-0
Molecular Formula: C32H22N6Na2O6S2
Formula Weight: 696.67
Odor: Odorless
Melting Point:
Formula:
C₃₂H₂₂N₆Na₂O₆S₂ MW: 696.67 g/mol Melting Pt: >360 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004028 CAS Number: 573-58-0 EINECS: 209-358-4 Merck Index: 14,2498 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...