1-Methylpyrrolidine 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
N-Methylpyrrolidine
B23799-36
B23799-22
AAB23799-36EA
148.18
USD
AAB23799-36
AAB23799-22
1-Methylpyrrolidine 98%
1-Methylpyrrolidine
Purity: 98%
CAS No.: 120-94-5
Molecular Formula: C5H11N
Formula Weight: 85.15
Flash Point: -18°(0°F)
Refractive Index: 1.4247
Odor: Amine-like
pH: 11.3
Melting Point: -90°
Boiling Point: 80-81°
UN No.: 3286
MDL No.: MFCD00003173
Formula:
C₅H₁₁N MW: 85.15 g/mol Boiling Pt: 80 Melting Pt: -76 Density: 0.799 Flash Pt: –18 °C (0 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003173 CAS Number: 120-94-5 EINECS: 204-438-5 UN: 3286 ADR: 3 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...