(±)-2-Aminoheptane 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2-Aminoheptane, (±)-Tuaminoheptane, 1-Methylhexylamine, 2-Heptylamine, (±)-1-Methylhexylamine, Tuaminoheptane
B22863-22
B22863-14
AAB22863-22EA
152
USD
AAB22863-22
AAB22863-14
(±)-2-Aminoheptane 98+%
(±)-2-Aminoheptane
Purity: 98+%
CAS No.: 123-82-0
Molecular Formula: C7H17N
Formula Weight: 115.22
Flash Point: 33°(91°F)
Refractive Index: 1.4190
Odor: Amine-like
pH: 11.45
Boiling Point: 142-144°
Beilstein Reference: 635676
Merck Reference: 14,9802
UN No.: 2733
MDL No.: MFCD00008101
Formula:
C₇H₁₇N MW: 115.22 g/mol Boiling Pt: 143 Density: 0.768 Flash Pt: 33 °C (91 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008101 CAS Number: 123-82-0 EINECS: 204-655-5 UN: 2734 ADR: 8 II Merck Index: 14,9802 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...