4'-Acetoxyacetophenone 99%

Supplier: Thermo Scientific Chemicals

Synonyms: 4-Acetylphenyl acetate, 4-Acetoxyacetophenone, p-Acetylphenyl acetate

B22284-14 B22284-22
AAAB22284-14EA 39.8 USD
AAAB22284-14 AAAB22284-22 AAAB22284-36
4'-Acetoxyacetophenone 99%
4'-Acetoxyacetophenone

Purity: 99% CAS No.: 13031-43-1 Molecular Formula: C10H10O3 Formula Weight: 178.19 Melting Point: 51-54° Boiling Point: 165-170°/14mm MDL No.: MFCD00017229

Formula: C₁₀H₁₀O₃
MW: 178.19 g/mol
Boiling Pt: 165…170 °C (14 mmHg)
Melting Pt: 51…54 °C
Storage Temperature: Ambient
MDL Number: MFCD00017229
CAS Number: 13031-43-1
EINECS: 235-894-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR