2,4,6-Tris(trifluoromethyl)-1,3,5-triazine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2,4,6-Tris(trifluoromethyl)-s-triazine
B22001-06
B22001-03
AAB22001-06EA
137
USD
AAB22001-06
AAB22001-03
2,4,6-Tris(trifluoromethyl)-1,3,5-triazine 98%
2,4,6-Tris(trifluoromethyl)-1,3,5-triazine
Purity: 98%
CAS No.: 368-66-1
Molecular Formula: C6F9N3
Formula Weight: 285.07
Flash Point: None
Refractive Index: 1.3220
Boiling Point: 95-96°
Beilstein Reference: 302339
MDL No.: MFCD00042436
Formula:
C₆F₉N₃ MW: 285.07 Boiling Pt: 95…96 °C Density: 1.596 Storage Temperature: Ambient |
MDL Number:
MFCD00042436 CAS Number: 368-66-1 EINECS: 206-709-3 UN: 2810 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...