Potassium chlorate 99+%

Supplier: Thermo Scientific Chemicals
Danger

A17075-30 A17075-0B A17075-0I
AAAA17075-30EA 53.93 USD
AAAA17075-30 AAAA17075-0B AAAA17075-0I
Potassium chlorate 99+%
Potassium chlorate

Purity: 99+% CAS No.: 3811-04-9 Molecular Formula: KClO3 Formula Weight: 122.55 Odor: Odorless Melting Point: 368° Boiling Point: 400° dec. Merck Reference: 14,7620 UN No.: 1485 MDL No.: MFCD00011361

Formula: KClO₃
Boiling Pt: 400 °C (dec.)
Melting Pt: 368 °C
Density: 2.32
Storage Temperature: Ambient
MDL Number: MFCD00011361
CAS Number: 3811-04-9
EINECS: 223-289-7
UN: 1485
ADR: 5.1,II
Merck Index: 14,7620

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR