Sodium hydrogen carbonate 99%

Supplier: Thermo Scientific Chemicals

A17005-0E A17005-0C A17005-36
AAAA17005-0EEA 81.66 USD
AAAA17005-0E AAAA17005-0C AAAA17005-36
Sodium hydrogen carbonate 99%
Sodium hydrogen carbonate

Purity: 99% CAS No.: 144-55-8 Molecular Formula: NaHCO3 Formula Weight: 84.01 Refractive Index: 1.500 Odor: Odorless pH: 8,1-8,4 Beilstein Reference: 4153970 Merck Reference: 14,8583 MDL No.: MFCD00003528

Formula: NaHCO₃
Melting Pt: 270 °C
Density: 2.159
Storage Temperature: Ambient
MDL Number: MFCD00003528
CAS Number: 144-55-8
EINECS: 205-633-8
Merck Index: 14,8583
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f6b1b80d2afb3deab0c4ca836a921fcaad637890&vid=1163b37d9cf1c10fe0ed252e02a51fe12d6b50dd&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR