n-Nonane 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A16177-AE
A16177-AP
AAAA16177-AEEA
112.54
USD
AAAA16177-AE
AAAA16177-AP
n-Nonane 99%
n-Nonane
Purity: 99%
CAS No.: 111-84-2
Molecular Formula: C9H20
Formula Weight: 128.26
Flash Point: 31°(87°F)
Refractive Index: 1.4054
Melting Point: -53°
Boiling Point: 150-152°
Beilstein Reference: 1696917
UN No.: 1920
MDL No.: MFCD00009574
Formula:
H₃C(CH₂)₇CH₃ Boiling Pt: 150…152 °C Melting Pt: –53 °C Density: 0.718 Flash Pt: 31 °C(87 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009574 CAS Number: 111-84-2 EINECS: 203-913-4 UN: 1920 ADR: "3",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...