2-Ethoxyethanol 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Ethyl glycol, Ethylene glycol monoethyl ether, Cellosolve
A16100-AP
A16100-0F
AAAA16100-APEA
53.83
USD
AAAA16100-AP
AAAA16100-0F
2-Ethoxyethanol 99%
2-Ethoxyethanol
A widely used solvent for nitrocelluloses and dyes
Purity: 99%
CAS No.: 110-80-5
Molecular Formula: C4H10O2
Formula Weight: 90.12
Flash Point: 44°(111°F)
Refractive Index: 1.4085
Odor: Ether-like
Melting Point: -90°
Boiling Point: 134-135°
Beilstein Reference: 1098271
Merck Reference: 14,3750
UN No.: 1171
MDL No.: MFCD00002869
Formula:
C₂H₅OCH₂CH₂OH MW: 90.12 g/mol Boiling Pt: 135 Melting Pt: -70 Density: 0.931 Flash Pt: 44 °C (111 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002869 CAS Number: 110-80-5 EINECS: 203-804-1 UN: 1171 ADR: 3 III REACH: 01-2119560582-38 Merck Index: 14,3750 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...