Potassium iodide 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Dipotassium diiodideKali iodide

A12704-0I A12704-30 A12704-0B A12704-0B A12704-0I A12704-30 A12704-18 A12704-18
AAAA12704-18EA 45.29 USD
AAAA12704-18 CAAAAA12704-0I CAAAAA12704-18 CAAAAA12704-30 AAAA12704-0I CAAAAA12704-0B AAAA12704-30 AAAA12704-0B
Potassium iodide 99%
Potassium iodide
Formula: Kl
MW: 166 g/mol
Boiling Pt: 1330 °C
Melting Pt: 681 °C
Density: 3.12
Storage Temperature: Ambient
MDL Number: MFCD00011405
CAS Number: 7681-11-0
EINECS: 231-659-4
Merck Index: 14,7643

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR