Potassium perchlorate 99%

Supplier: Thermo Scientific Chemicals
Danger

A11296-36 A11296-22 A11296-0E
AAAA11296-36EA 128.59 USD
AAAA11296-36 AAAA11296-22 AAAA11296-0E
Potassium perchlorate 99%
Potassium perchlorate

Purity: 99% CAS No.: 7778-74-7 Molecular Formula: KClO4 Formula Weight: 138.55 Odor: Odorless Melting Point: 301-304° UN No.: 1489 MDL No.: MFCD00011362

Formula: ClKO₄
MW: 138.55 g/mol
Melting Pt: 301…304 °C
Density: 2.52
Storage Temperature: Ambient
MDL Number: MFCD00011362
CAS Number: 7778-74-7
EINECS: 231-912-9
UN: 1489
ADR: "5.1",II
Merck Index: 14,7653

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR