Biphenyl (PCB No. 0) 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
MDL: MFCD00003054
Beilstein: 1634058
Soluble in alcohol and ether
Purity: 99%
CAS No.: 92-52-4
Molecular Formula: C12H10
Formula Weight: 154.21
Flash Point: 113°(235°F)
Odor: Pleasant
Boiling Point: 254-255°
Solubility: Soluble in alcohol and ether
Beilstein Reference: 1634058
Merck Reference: 14,3314
UN No.: 3077
MDL No.: MFCD00003054
Formula:
C₁₂H₁₀ Boiling Pt: 254…255 °C Melting Pt: 69…72 °C Density: 0.990 Flash Pt: 113 °C (235 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003054 CAS Number: 92-52-4 EINECS: 202-163-5 UN: 3077 ADR: 9,III Merck Index: 14,3314 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...