Molybdenum ≥99.9% (metals basis), wire, hard, Diameter 0.203 mm (0.008 in)
Supplier: Thermo Scientific Chemicals
045134-G1
045134-G6
045134-G5
AA45134-G1EA
43.4
USD
AA45134-G1
AA45134-G6
AA45134-G5
Molybdenum ≥99.9% (metals basis), wire, hard, Diameter 0.203 mm (0.008 in)
Molybdenum
Purity: 99.9%
CAS No.: 7439-98-7
Physical Form: ≈0.33g/m
MDL No.: MFCD00003465
Formula:
Mo Boiling Pt: 3617 °C (1013 hPa) Melting Pt: 2622 °C Density: 10.28 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003465 CAS Number: 7439-98-7 EINECS: 231-107-2 Merck Index: 12,06317 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...