Bismuth ≥99.9% (metals basis), ingot/button, Diameter ∼36 mm (1.4 in), Thickness 8 mm (0.3 in)

Supplier: Thermo Scientific Chemicals

45102-KF
AA45102-KFEA 80.15 USD
AA45102-KF
Bismuth ≥99.9% (metals basis), ingot/button, Diameter ∼36 mm (1.4 in), Thickness 8 mm (0.3 in)
Bismuth
Formula: Bi
MW: 208.98 g/mol
Boiling Pt: 1490 °C (1013 hPa)
Melting Pt: 271…272 °C
Density: 9.78 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134033
CAS Number: 7440-69-9
EINECS: 231-177-4
Merck Index: 12,01297

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR