Nickel, gauze woven from wire 60 mesh, Diameter 0.18 mm (0.007 in)
Supplier: Thermo Scientific Chemicals
|
Danger
|
045099-FL
045099-HB
045099-GT
AA45099-FLEA
75.53
USD
AA45099-FL
AA45099-HB
AA45099-GT
Nickel, gauze woven from wire 60 mesh, Diameter 0.18 mm (0.007 in)
Nickel
CAS No.: 7440-02-0
Physical Form: Wire Cloth
Odor: Odorless
MDL No.: MFCD00011137
Formula:
Ni MW: 58.69 g/mol Boiling Pt: 3007 Melting Pt: 1453 Density: 8.9 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011137 CAS Number: 7440-02-0 EINECS: 231-111-4 UN: 3089 ADR: 4.1 III Merck Index: 12,06582 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...