Iron ≥99.99% (metals basis), pieces, irregularly shaped 3.2-6.4 mm (0.125-0.25 in)
Supplier: Thermo Scientific Chemicals
045090-A1
045090-22
045090-36
AA45090-A1EA
332
USD
AA45090-A1
AA45090-22
AA45090-36
Iron ≥99.99% (metals basis), pieces, irregularly shaped 3.2-6.4 mm (0.125-0.25 in)
Iron
Purity: 99.99%
CAS No.: 7439-89-6
Appearance: Grey
MDL No.: MFCD00010999
Formula:
Fe MW: 55.85 g/mol Boiling Pt: 3000 Melting Pt: 1535 Density: 7.86 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010999 CAS Number: 7439-89-6 EINECS: 231-096-4 Merck Index: 13,05109 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...