Zirconium ≥99.2% (metals basis excluding Hf), wire (max. 4.5% Hf), annealed, Diameter 0.5 mm (0.020 in)

Supplier: Thermo Scientific Chemicals

045056-G5 045056-G1
AA45056-G5EA 92 USD
AA45056-G5 AA45056-G1
Zirconium ≥99.2% (metals basis excluding Hf), wire (max. 4.5% Hf), annealed, Diameter 0.5 mm (0.020 in)
Zirconium

Purity: 99.2% CAS No.: 7440-67-7 Physical Form: ≈1.31g/m Odor: Odorless MDL No.: MFCD00011303

Formula: Zr
MW: 91.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011303
CAS Number: 7440-67-7
EINECS: 231-176-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR