Molybdenum ≥99.9% (metals basis), wire, annealed, Diameter 1.58 mm (0.0625 in)

Supplier: Thermo Scientific Chemicals

45055-G5 45055-G6
AA45055-G6EA 380.99 USD
AA45055-G6 AA45055-G5
Molybdenum ≥99.9% (metals basis), wire, annealed, Diameter 1.58 mm (0.0625 in)
Molybdenum
Formula: Mo
MW: 95.94 g/mol
Boiling Pt: 3617 °C (1013 hPa)
Melting Pt: 2622 °C
Density: 10.28 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003465
CAS Number: 7439-98-7
EINECS: 231-107-2
Merck Index: 12,06317

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR