Molybdenum ≥99.9% (metals basis), wire, annealed, Diameter 1.14 mm (0.045 in)

Supplier: Thermo Scientific Chemicals

045054-G6 045054-G5 045054-G9
AA45054-G6EA 266 USD
AA45054-G6 AA45054-G5 AA45054-G9
Molybdenum ≥99.9% (metals basis), wire, annealed, Diameter 1.14 mm (0.045 in)
Molybdenum

Purity: 99.9% CAS No.: 7439-98-7 Physical Form: ≈10.5g/m MDL No.: MFCD00003465

Formula: Mo
Boiling Pt: 3617 °C (1013 hPa)
Melting Pt: 2622 °C
Density: 10.28 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003465
CAS Number: 7439-98-7
EINECS: 231-107-2
Merck Index: 12,06317

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR