Iron ≥99.95% (metals basis), rods, Diameter 7.62 mm (0.30 in)

Supplier: Thermo Scientific Chemicals

44918-DQ
AA44918-DQEA 157.99 USD
AA44918-DQ
Iron ≥99.95% (metals basis), rods, Diameter 7.62 mm (0.30 in)
Iron
Formula: Fe
MW: 55.85 g/mol
Boiling Pt: 3000 °C (1013 hPa)
Melting Pt: 1535 °C
Density: 7.86 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010999
CAS Number: 7439-89-6
EINECS: 231-086-4
Merck Index: 13,05109

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR