Cobalt ≥99.95% (metals basis), wire, Diameter 1.5 mm (0.06 in)
Supplier: Thermo Scientific Chemicals
|
Danger
|
44524-BU
44524-BQ
AA44524-BQEA
72.09
USD
AA44524-BQ
AA44524-BU
Cobalt ≥99.95% (metals basis), wire, Diameter 1.5 mm (0.06 in)
Cobalt
Formula:
Co MW: 58.93 g/mol Boiling Pt: 2870 °C (1013 hPa) Melting Pt: 1495 °C Density: 8,9 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010935 CAS Number: 7440-48-4 EINECS: 231-158-0 Merck Index: 12,02488 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...