Cobalt ≥99.95% (metals basis), foil, Thickness 1.5 mm (0.06 in)

Supplier: Thermo Scientific Chemicals
Danger

44523-FF 44523-FI
AA44523-FFEA 120.99 USD
AA44523-FF AA44523-FI
Cobalt ≥99.95% (metals basis), foil, Thickness 1.5 mm (0.06 in)
Cobalt
Formula: Co
MW: 58.93 g/mol
Boiling Pt: 2870 °C (1013 hPa)
Melting Pt: 1495 °C
Density: 8,9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010935
CAS Number: 7440-48-4
EINECS: 231-158-0
Merck Index: 12,02488

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR