Tetrahydrofuran ≥99.8% unstabilised, Biograde for DNA and peptides synthesis

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Oxolane, 1,4-epoxy-Butane, Butylene oxide, Butane alpha,delta-oxide, Tetramethylene oxide, THF, Oxacyclopentane, Cyclotetramethylene oxide, Diethylene oxide

044505-K2 044505-K7
AAA44505-K2EA 96.3 USD
AAA44505-K2 AAA44505-K7
Tetrahydrofuran ≥99.8% unstabilised, Biograde for DNA and peptides synthesis
Tetrahydrofuran

Applications: Nucleic acid and peptide synthesis


Purity: 99.8% CAS No.: 109-99-9 Molecular Formula: C4H8O Formula Weight: 72.11 Flash Point: -17°(1°F) Physical Form: Liquid Density: 0.889 Refractive Index: 1.4070 Appearance: Colorless Odor: Ether-like Melting Point: -108° Boiling Point: 66° Beilstein Reference: 102391 Merck Reference: 14,9211 UN No.: 2056 MDL No.: MFCD00005356


Nucleic acid and peptide synthesis

Formula: C₄H₈O
MW: 72.11 g/mol
Boiling Pt: 66
Melting Pt: -108.5
Density: 0.889
Flash Pt: –17 °C (1 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005356
CAS Number: 109-99-9
EINECS: 203-726-8
UN: 2056
ADR: 3 II
REACH: 05-2114615188-48
Merck Index: 14,9211
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b2d7611caf0127fbef941994edd1d7e69487d54a&vid=2a64e21bd2e13636c3038adf839b3e9ff18509c5&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR