Aluminium ≥99.999% (metals basis), foil, Thickness 0.025 mm (0.001 in)
Supplier: Thermo Scientific Chemicals
Synonyms:
Aluminum
044233-HC
044233-GA
AA44233-HCEA
400
USD
AA44233-HC
AA44233-GA
Aluminium ≥99.999% (metals basis), foil, Thickness 0.025 mm (0.001 in)
Aluminium
Purity: 99.999%
CAS No.: 7429-90-5
Physical Form: ≈0.675g/50x200mm
Odor: Odorless
MDL No.: MFCD00134029
Formula:
Al MW: 26.98 g/mol Boiling Pt: 2327 Melting Pt: 660 Density: 2.7 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00134029 CAS Number: 7429-90-5 EINECS: 231-072-3 Merck Index: 13,00321 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...