Copper ≥99.99% (metals basis), rods, Diameter 2.0 mm (0.08 in)

Supplier: Thermo Scientific Chemicals

043973-BS 043973-BV
AA43973-BSEA 39.9 USD
AA43973-BS AA43973-BV
Copper ≥99.99% (metals basis), rods, Diameter 2.0 mm (0.08 in)
Copper

Purity: 99.99% CAS No.: 7440-50-8 Physical Form: ≈2.81g/10cm Odor: Odorless MDL No.: MFCD00010965

Formula: Cu
Boiling Pt: 2595 °C (1013 hPa)
Melting Pt: 1083 °C
Density: 8.94 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010965
CAS Number: 7440-50-8
EINECS: 231-159-6
Merck Index: 13,02545

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR