Titanium, gauze woven from wire 18 mesh, Diameter 0.28 mm (0.011 in)
Supplier: Thermo Scientific Chemicals
|
Warning
|
43794-GN
43794-GY
AA43794-GNEA
120.99
USD
AA43794-GN
AA43794-GY
Titanium, gauze woven from wire 18 mesh, Diameter 0.28 mm (0.011 in)
Titanium
Formula:
Ti MW: 47.88 g/mol Boiling Pt: 3262 °C (1013 hPa) Melting Pt: 1660 °C Density: 4,51 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011264 CAS Number: 7440-32-6 EINECS: 231-142-3 Merck Index: 12,09610 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...