Malonic acid ≥99.5%, Reagent Grade

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1,3-Propanedioic acid

031715-22 031715-A4 031715-36
AAA31715-22EA 68.94 USD
AAA31715-22 AAA31715-A4 AAA31715-36
Malonic acid ≥99.5%, Reagent Grade
Malonic acid
Formula: C₃H₄O₄
MW: 104.06 g/mol
Boiling Pt: 387 °C (1013 hPa)
Melting Pt: 132…136 °C (dec.)
Density: 1.62
Flash Pt: 157 °C (314 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002707
CAS Number: 141-82-2
EINECS: 205-503-0
Merck Index: 13,05732

Order Now

Specification Test Results

Form Crystalline

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR