Iodine 99,99 Metals basis, Crystalline

Supplier: Thermo Scientific Chemicals
Warning

000158-18 000158-30 000158-22
AAA00158-18EA 85.54 USD
AAA00158-18 AAA00158-30 AA00158-22
Iodine 99,99 Metals basis, Crystalline
Iodine
Formula: I₂
MW: 253.81 g/mol
Boiling Pt: 184.3
Melting Pt: 114 (subl.)
Density: 4.930
Storage Temperature: Ambient
MDL Number: MFCD00011355
CAS Number: 7553-56-2
EINECS: 231-442-4
UN: 3495
ADR: 8 III
Merck Index: 13,05036

Order Now

Specification Test Results

Form Solid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR