Sodium periodate

Supplier: G-Biosciences
Danger

Synonyms: Sodium metaperiodate

BKC-15 BKC-15
82021-448EA 50.92 USD
82021-448 CA82021-448
Sodium periodate
Sodium periodate

G-Biosciences' Sodium meta -Periodate is a mild oxidizing agent that converts carbohydrates to activated active aldehydes


G-Biosciences' Sodium meta-Periodate is also suitable for oxidizing glycoproteins.


It is used in coupling to amines with cyanborohydride reduction.

Formula: NaIO₄
MW: 213.89 g/mol
Boiling Pt: 300 °C (1013 hPa)
Melting Pt: 175 °C
Density: 3.87 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003534
CAS Number: 7790-28-5
EINECS: 232-197-6
UN: 1479
ADR: 5.1,II
Merck Index: 13,08714

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR