Nunc CryoTube Colored Caps, Thermo Scientific
Supplier: Thermo Scientific
336094 363436 336088 336092 336081
73521-223CS 1333.68 USD
73521-223 73521-394 73521-221 73521-220 73521-222
Nunc CryoTube Colored Caps, Thermo Scientific
Closures Test/Sample Tube Closures
Nunc CryoTube colored caps are replacement polypropylene screw caps with a silicone gasket to provide the best seal
They are made to be used with Nunc internally threaded CryoTube vials.