sensION+ ORP Electrodes, Hach
Supplier: Hach
sensION+
LZW5056970002 LZW5057970002 LZW5055970002
89174-202EA 769.23 USD
89174-202 89174-204 89174-226
sensION+ ORP Electrodes, Hach
Electrodes Redox Electrodes
sensION+ ORP electrodes have a mill volt range of ± 2000 mV and are equipped with a platinum wire
sensION+ ORP electrodes can be used in a variety of applications such as oxide-reduction titrations, galvanic baths, general applications, etc.
ORP probes for the sensION+ have several common features, such as a ceramic diaphragm and a glass body.
Certifications: .CE.WEEE.