L(-)-Menthol

Supplier: USP
Warning

Synonyms: (-)-MentholLevomenthol(1R,2S,5R)-2-Isopropyl-5-methylcyclohexanol(1R)-(-)-MentholL(-)-Menthyl alcoholol(1R,2S,5R)-(-)-Menthol(1α,2β,4β)-(-)-2-Hydroxy-1-isopropyl-4-methylcyclohexane(1S,2R,4R)-(-)-2-Hydroxy-1-isopropyl-4-methylcyclohexane

1381709
101095-420EA 429.23 USD
101095-420
L(-)-Menthol
L(-)-Menthol
Formula: C₁₀H₂₀O
MW: 156.27 g/mol
Boiling Pt: 212 °C (1013 hPa)
Melting Pt: 41…44 °C
Density: 0,89 g/cm³ (20 °C)
Flash Pt: 96 °C
Storage Temperature: Ambient
MDL Number: MFCD00062979
CAS Number: 2216-51-5
EINECS: 218-690-9
Merck Index: 13,05861

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR