L(+)-Lysine monohydrochloride

Supplier: USP

Synonyms: (S)-(+)-2,6-Diaminohexanoic acid monohydrochlorideH-Lys-OH.HClL-Lysine monohydrochlorideH-Lys-OH·HClL-Lysine monohydrocholoride(S)-(+)-Lysine monohydrochlorideLysine HydrochlorideL-Lysine Hydrochloride(S)-(+)-2,6-Diaminocaproic acid monohydrochloride

1372005
101095-352EA 513.85 USD
101095-352
L(+)-Lysine monohydrochloride
L(+)-Lysine monohydrochloride
Formula: H₂N(CH₂)₄CH(NH₂)CO₂H·HCl
MW: 182.65 g/mol
Melting Pt: 263…264 °C
Storage Temperature: Ambient
MDL Number: MFCD00064564
CAS Number: 657-27-2
EINECS: 211-519-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR